missing translation for 'onlineSavingsMsg'
Learn More

Clathrin Heavy Chain 2/CHC22/CLTCL1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£181.00 - £364.00

Specifications

Antigen Clathrin Heavy Chain 2/CHC22/CLTCL1
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18663191
View Documents
Novus Biologicals
NBP2-92400-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18635470
View Documents
Novus Biologicals
NBP2-92400-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Clathrin Heavy Chain 2/CHC22/CLTCL1 Polyclonal antibody specifically detects Clathrin Heavy Chain 2/CHC22/CLTCL1 in Human samples. It is validated for Western Blot
Specifications

Specifications

Clathrin Heavy Chain 2/CHC22/CLTCL1
Western Blot
Unconjugated
Rabbit
B Cell Development and Differentiation Markers, Immunology, Neurodegeneration, Neuroscience
PBS with 50% glycerol, pH7.3.
8218
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
CHC22, clathrin heavy chain 2, clathrin heavy chain on chromosome 22, clathrin, heavy chain-like 1, Clathrin, heavy polypeptide D, CLTCL, heavy polypeptide-like 1
A synthetic peptide corresponding to a sequence within amino acids 1600-1663 of human Clathrin Heavy Chain 2/CHC22/CLTCL1 (NP_009029.3). IQVMREYLSKVDKLDALESLRKQEEHVTEPAPLVFDFDGHE
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.