missing translation for 'onlineSavingsMsg'
Learn More

Clathrin Heavy Chain 1/CHC17 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18376414 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376414 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376414 Supplier Novus Biologicals Supplier No. NBP299436AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Clathrin Heavy Chain 1/CHC17 Polyclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin Heavy Chain 1/CHC17
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse Clathrin Heavy Chain 1/CHC17.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 1213
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.