missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Classic Rack for Chest Freezers, 50 mm Box Height, with Boxes
Classic Rack is suitable for use with all chest freezer models.
Brand: Tenak™ TE21456B
Additional Details : Weight : 3.00000kg
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Stainless Steel | |
140 x 140 x 665 mm |
Boxes (12) | |
Chest Frseezers | |
50 mm (Box) |