missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Classic Children's Shoecovers with Plastic Sole

Product Code. 17720099 Shop All Medicom Products
Change view
Click to view available options
Classic Children's Shoecovers with Plastic Sole; Color: White, Blue; Size: Small; For Use With (Application): Medical; Material: Polypropylene, Polyethylene; Thickness: 50 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17720099 Classic Children's Shoecovers with Plastic Sole; Color: White, Blue; Size: Small; For Use With (Application): Medical; Material: Polypropylene, Polyethylene; Thickness: 50 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17720099 Supplier Kolmi Hopen™ Supplier No. 10.012E

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

White, children's shoecovers with skid-resistant soles, ideal for the medical setting.

  • Child model suitable for infants (1 to 4 years old)
  • Anti-slip sole reduces the risk of falling
  • Polypropylene with polyethylene sole
  • Classic style
  • Welded and elasticated

Specifications

Certifications/Compliance ISO 9001
Color White, Blue
Dimensions (L x W) 23 x 12 cm
For Use With (Application) Medical
Material Polypropylene, Polyethylene
Size Small
Packaging Quantity 600 Pk.
Sterility Non-sterile
Thickness 50 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.