Learn More
Abeba™ Classic 31001 Shoes
Product Code. p-7147448
Click to view available options
Size:
36
37
38
39
40
41
42
43
45
46
Unit Size:
1 pair
Product Code. 15415714
Brand: Abeba™ 31001/36
Only null left
Add to basket
Add to basket
Animal fats, oil and fuel resistant. Leather/microfiber construction.
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
  • Comfortable shape
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E, FO SRC
Color White
Liner Material Microfiber
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 31001
Toe Material Steel
Resistant To Animal fats, oil and fuel