Learn More
Abeba™ Classic 1001 Shoes
Product Code. 15656567
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. 15656567
Brand: Abeba™ 1001/36
This item requires a price update prior to being ordered.
Request a Price for this item here.
Animal fats, oil and fuel resistant. Leather/microfiber construction.
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E, FO SRC
Color White
Liner Material Microfiber
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1001
Toe Material Steel
Resistant To Animal fats, oil and fuel