Learn More
Abeba™ Classic 1000 Shoes
Product Code. 15696557
Click to view available options
Size:
36
42
43
44
45
46
Unit Size:
1 pair
Product Code. 15696557
Brand: Abeba™ 1000/36
Only null left
Add to basket
Add to basket
Animal fats, oil and fuel resistant. Leather/microfiber construction.
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Grained leather
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • Buckle fastening
  • Heel patch microfiber
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E, FO SRC
Color White
Liner Material Microfiber
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1000
Toe Material Steel
Resistant To Animal fats, oil and fuel