missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M2 Weight Set

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M2 Weights are ideal for the verification or calibration of class M3 weights.

Brand:  Zwiebel™ PBI5683C

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11830942

  • £285.14 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Class M2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications

    Specifications

    1 to 100 g
    For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers