Learn More
Zwiebel™ Class M2 Chrome Weight Set
Product Code. 11724714
Change view
Click to view available options
Mass:
1 to 1000 g
1 to 200 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass unitSize
11724714 1 to 1000 g Each
11704714 1 to 200 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11724714 Supplier Zwiebel™ Supplier No. PBI23705
Only null left
Add to basket
Add to basket
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M2 Weights are ideal for the verification or calibration of class M3 weights.
  • Class M2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Mass 1 to 1000 g