Learn More
Zwiebel™ Class M2 Chrome Weight Set
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M2 Weights are ideal for the verification or calibration of class M3 weights.
Brand:  Zwiebel™ PBI23703
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11704714
  • £329.66 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
  • Class M2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications
    Specifications
    1 to 200 g