missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Weights with Certificate

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.

Brand:  Zwiebel™ PBI23724

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11784744

  • £292.56 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

  • Class M1 calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications

Specifications

Cast Iron
20 kg
±1000 mg
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers