missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class F1 Weights

Product Code. p-8006535
Click to view available options
Mass:
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11710623

Brand: Zwiebel™ PBI2301T

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use with weighing instruments of medium accuracy class II. Zwiebel™ Class F1 Box Weights are ideal for the verification or calibration of class F2 weights.

  • Class F1 weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box

Specifications

Mass 100 mg
Tolerance ±0.05 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.