Learn More
Zwiebel™ Class F1 Weight Set
Brand:  Zwiebel™ PBI23143
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11790803
  • £1425.70 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Specifications
1 to 50 g
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II