Learn More
Zwiebel™ Class F1 Weight Set
Brand: Zwiebel™ PBI23158
Code : A0
Additional Details : Weight : 0.80000kg
Specifications
1 to 500 mg | |
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |