Learn More
Zwiebel™ Class F1 Weight Box with Certificate
Brand:  Zwiebel™ PBI56819
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11870892
  • £389.02 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Specifications
100 g
±5 mg
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II