Learn More
Zwiebel™ Class F1 Box Weights with Certificate
Use with weighing instruments of special accuracy class I and high accuracy class II. Zwiebel™ Class F1 Weights are intended for use in the verification or calibration of class F2 weights.
£152.64 - £1717.20
Specifications
For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |
---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | |||||
11770803
|
Zwiebel™
PBI23141 |
±50 mg |
£1717.20
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11780793
|
Zwiebel™
PBI23138 |
±5.0 mg |
£483.36
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11750793
|
Zwiebel™
PBI23135 |
±0.50 mg |
£202.46
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11720793
|
Zwiebel™
PBI23132 |
±0.20 mg |
£169.60
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11760773
|
Zwiebel™
PBI23129 |
±0.1 mg |
£152.64
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
11730793
|
Zwiebel™
PBI23133 |
±0.25 mg |
£174.90
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11760803
|
Zwiebel™
PBI23140 |
±25 mg |
£1102.40
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11740793
|
Zwiebel™
PBI23134 |
±0.30 mg |
£187.62
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Specifications
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |