Learn More
Zwiebel™ Class F1 Box Weights
Use with weighing instruments of medium accuracy class II. Zwiebel™ Class F1 Box Weights are ideal for the verification or calibration of class F2 weights.
Brand:  Zwiebel™ PBI2301H
Code : A0
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12021633
  • £53.00 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications
    Specifications
    200 mg
    ±0.06 mg
    For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II