missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class E2 Weight with COFRAC Calibration Certificate
Description
- Class E2 calibration weight
- In compliance with OIML R111
- Delivered with a COFRAC calibration certificate guaranteeing traceability
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Mass | 500 mg |
| Tolerance | ±0.025 mg |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?