Learn More
Zwiebel™ Class E2 Weight Box with COFRAC Calibration Certificate
Brand:  Zwiebel™ PBI56814
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11830892
  • £405.98 / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Specifications
500 g
±0.8 mg
For use with Weighing Instruments of Special Accuracy Class I