Learn More
Zwiebel™ Class E2 Box Weight
Specifications
Specifications
| Mass | 10 g |
| Tolerance | ±0.06 mg |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |