missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate
Description
- Class E2 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Mass | 50 g |
| Tolerance | ±0.1 mg |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?