missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate

Product Code. 11715193
Change view
Click to view available options
Mass:
1 g
1 to 100 g
10 g
100 g
2 g
20 g
200 g
5 g
5 kg
50 g
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass unitSize
11715193 100 g Each
11710263 1 to 100 g Each
11725193 200 g Each
11785183 10 g Each
11755183 1 g Each
11795183 20 g Each
11765183 2 g Each
11775183 5 g Each
11705193 50 g Each
11745203 5 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 11715193 Supplier Zwiebel™ Supplier No. PBI23878
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Calibrate weights of lower accuracy classes and are used with class I weighing instruments. Zwiebel™ Class E2 Box Weights are intended for use with class F1 weights.

  • Class E2 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box

Specifications

Mass 100 g
Tolerance ±0.16 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.