Learn More
VITLAB™ Class B SAN Short Form Measuring Cylinders
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B SAN Short Form Measuring Cylinders feature easy to read, raised scale and ring marks at the primary scale points.
Brand:  VITLAB™ 64191
Product Code. 10578242
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Crystal clear graduated cylinder
  • Calibrated (In)
  • Class B tolerances according to DIN 12681 / ISO 6706.
  • Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded