Learn More
VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale feature easy to read, raised, embossed blue scale and ring marks at the primary scale points.
Brand: VITLAB™ 652081
Additional Details : Weight : 0.01000kg
Description
- Highly transparent polypropylene graduated cylinder
- Hexagonal base with bottom studs provides high stability
- Calibrated (In)
- Class B tolerances according to DIN 12681 / ISO 6706.
- Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
- To preserve markings, do not clean at temperatures exceeding 60°C