missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Class B PMP Volumetric Flask

Autoclaving up to 121°C will not cause permanently exceeded error limits. Cleaning up to max. 60°C is recommended to preserve marks and inscriptions.

Brand:  BRAND™ 36148

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 17420144

  • £35.15 / Pack of 2
In Stock
to see stock.
Add to basket

Description

Description

  • Transparent PMP (Polymethylpentene) construction
  • Includes polypropylene stopper
  • Error limits according to class B, DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Exposure to temperatures up to 121°C (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60°C is recommended to preserve marks and inscriptions
  • Height = Height without stopper
Specifications

Specifications

DIN EN ISO 1042
250 mL
225 mm
1
±0.30 mL
B
Polypropylene
PMP
±0.30 mL
121°C
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers