missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Class A USP Brown Graduated Measuring Pipettes

Provided with a USP dated batch identification certificate for uniformity.

Brand:  Hirschmann™ 110017027

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11701526

  • £86.25 / Pack of 6
Estimated Shipment: 30-08-2024
to see stock.
Add to basket

Description

Description

  • With silver diffusion ink for permanent markings
  • Brown graduation marks
  • Main point ring graduation
  • Graduated to tip
  • Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
  • Available in a range of volumes
Specifications

Specifications

USP
AR-GLAS
Yes
0.2 mL
25 mL
Measuring Pipet
A
Volumetric accuracy in pipetting
Brown
0.05 mL
450 ±5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers