missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Class A Measuring Cylinders, Schellbach Graduation

Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Schellbach Graduation are manufactured from durable DURAN™ glass tubing.

Brand:  Hirschmann Instruments™ 2220160

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10238283

  • £18.20 / Pack of 2


Estimated Shipment
23-05-2018
Add to basket

Description

Description

  • With spout, main point ring graduation
  • Blue, Schellbach stain graduation
  • Conformity certified, with dated batch certificate
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

140mm
DIN EN ISO 4788
10mL
0.2mL
Glass (Untyped)
A
0.1
0.1
Videos