Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper
Product Code. 12857342 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
2 mL
100 mL
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12857342 100 mL Pack of 2
16357987 2 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12857342 Supplier BRAND™ Supplier No. 237249
Only null left
Add to basket
Add to basket
Borosilicate volumetric flask with polypropylene stopper and ISO single batch certificate.
  • Borosilicate 3.3
  • DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Includes ISO certificate single batch delivery
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO Single Batch
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat