Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper
Product Code. 16367987 Shop All Brand Products
Change view
Click to view available options
:
Volumetric Flask
Product Code.
16367987 Volumetric Flask
1 options
Product Code. 16367987 Supplier BRAND™ Supplier No. 236953
Only null left
Add to basket
Add to basket
Borosilicate volumetric flask with polypropylene stopper and USP single batch certificate.
  • Borosilicate 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Includes USP certificate single batch delivery
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1 L
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat