Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper
Product Code. 30564695 Shop All Brand Products
Change view
Click to view available options
Socket Joint:
NS 12/21
NS 14/23
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Socket Joint unitSize
30564695 NS 12/21 Pack of 2
17716444 NS 12/21 Each
10111145 NS 14/23 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30564695 Supplier BRAND™ Supplier No. 937249
  • ONLINE EXCLUSIVE PRICE
    £96.79
    £87.82 / Pack of 2
    Save £8.97
    9% Off
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Borosilicate volumetric flask with polypropylene stopper and ISO individual certificate.
  • Borosilicate 3.3
  • DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Includes ISO individual certificate
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO Individual
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Socket Joint NS 12/21
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat