missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Volumetric Flask with Single Certificate

Product Code. 12353986
Click to view available options
Capacity (Metric):
25 mL
100 mL
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 12353986

Brand: Poulten Graf™ RB258/AM/WAC/E/5

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Created from borosilicate glass. Poulten Graf™ FORTUNA™ Borosilicate Glass Class A Volumetric Flasks offers excellent chemical resistance and are best for obtaining a precise measurement of chemical solutions.

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Single Certificate are individually numbered for identification and traceability.

  • Polypropylene stopper included
  • Available in amber or clear glass
  • ISO and ASTM certified
  • Single mark gradation

Specifications

Certifications/Compliance ISO and ASTM certified
Color Amber
Capacity (Metric) 25 mL
Material Borosilicate Glass
No. per Case 5
Graduated Yes
Class A
Closure Type Polystop Stopper
Closure Size 10/19
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.