missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Volumetric Flask with Certificate of Calibration

Product Code. 10356152
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
Color:
Amber
Clear
Unit Size:
Each
Pack of 2
Pack of 5
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color unitSize
10356152 200 mL Amber Each
10191422 1000 mL Clear Pack of 2
10683283 100 mL Clear Pack of 5
10529672 100 mL Amber Pack of 5
10663093 10 mL Clear Pack of 5
12393906 15 mL Clear Pack of 5
12343946 2000 mL Amber Each
12383916 2000 mL Clear Each
10531563 200 mL Clear Pack of 2
10102662 250 mL Amber Pack of 2
10465782 20 mL Clear Pack of 5
10603673 250 mL Clear Pack of 2
10131382 25 mL Clear Pack of 5
12393916 5 L Clear Each
10151332 5 mL Clear Pack of 5
10529482 50 mL Amber Pack of 5
10521563 50 mL Clear Pack of 5
10263952 500 mL Clear Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 10356152 Supplier Poulten Graf™ Supplier No. US258/AM/WAC/H/2

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

  • Tooled and ground neck fitted with a Polystop™ polypropylene interchangeable stopper
  • Single mark gradation
  • Available in amber or clear glass
  • Each flask carries an individual number for identification and traceability

Specifications

Certifications/Compliance USP, ISO 1042, DIN 12664
Closure Color Red
Closure Material Polyethylene
Color Amber
Capacity (Metric) 200 mL
Material Borosilicate Glass
No. per Case 2
Tolerance 0.10 mL
Graduated Yes
Class A
Closure Size 14.5/23
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.