missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Trapezoidal Volumetric Flask

Created from borosilicate glass. Poulten Graf™ Volumetric Flasks with Class A Stopper are used to prepare standard solutions.

Brand:  Poulten Graf™ RB257/AM/C/5

Code : 01

Additional Details : Weight : 0.11844kg

 View more versions of this product

Product Code. 12393976

  • £149.64 / Pack of 5
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

  • Polypropylene stopper included
  • Each flask individually numbered for identification and traceability
  • Volac™ borosilicate glass
Specifications

Specifications

Polypropylene
3 mL
5
Yes
Trapezoidal
Amber
Borosilicate Glass
±0.025 mL
A
7.5/16
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers