missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Trapezoidal Volumetric Flask

Created from borosilicate glass. Poulten Graf™ Volumetric Flasks with Class A Stopper are used to prepare standard solutions.

Brand:  Poulten Graf™ RB257/E/5

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12363976

  • £132.10 / Pack of 5
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

  • Polypropylene stopper included
  • Each flask individually numbered for identification and traceability
  • Volac™ borosilicate glass
Specifications

Specifications

Polypropylene
5 mL
5
Yes
Trapezoidal
Clear
Borosilicate Glass
±0.025 mL
A
7/16
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers