missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Class A Amber Glass White Graduated Measuring Flasks

Product Code. 13234663
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
Unit Size:
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
13234663 10 mL Pack of 2
10717803 25 mL Pack of 2
12739886 50 mL Pack of 2
10727803 100 mL Pack of 2
10652364 200 mL Pack of 2
10776253 250 mL Pack of 2
10597383 500 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 13234663 Supplier Hirschmann™ Supplier No. 264016127
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provided in amber glass for the protection of light-sensitive contents.

  • White graduation
  • Class A
  • With dated batch identification
  • Standard ground and PE stopper
  • Manufactured from DURAN™ borosilicate glass for high chemical and thermal resistance
  • USP certified

Specifications

Closure Material Polyethylene
Color Amber
Capacity (Metric) 10 mL
Material Borosilicate Glass
For Use With (Application) Liquid handling of light-sensitive products
Height (Metric) 90 ± 5 mm
Standard Taper Joint Ground 10/19
Graduated Yes
Class A
Bottom Shape Flat
Closure Type Stopper
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.