missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks

Product Code. 11726742
This item is not returnable. View return policy
Product Code. 11726742 Supplier Hirschmann Instruments™ Supplier No. 2640286
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks are manufactured from durable DURAN™ glass tubing.

  • Amber glass
  • Acc. to DIN EN ISO 1042
  • With NS ground and hollow amber glass stopper
  • Conformity certified, with dated batch identification

Specifications

Closure Color Amber
Color Amber
Capacity (Metric) 250 mL
Material Glass
Graduations Yes
Standard Taper Joint 14/23
Tolerance 0.15
Graduated Yes
Class A
Closure Type Glass Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.