Learn More
BRAND™ Class A Amber Borosilicate Volumetric Flask with Glass Stopper
Product Code. p-200104818 Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
1 L
Certifications/Compliance:
USP Single
USP Single Batch
Unit Size:
Each
Pack of 2
Product Code. 16357997
Brand: BRAND™ 237464
Only null left
Add to basket
Add to basket
Amber volumetric flask with glass stopper and USP certificate.
  • Borosilicate 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Includes USP certificate, single, single batch, or batch delivery
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Contain
Certifications/Compliance USP Single Batch
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat