Learn More
Manutan Class 8 corrosive IATA/ICAO labels,100mm x 100mm
Product Code. 12868958
Encompass_Preferred
Product Code. 12868958 Supplier Manutan Supplier No. A352642
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A range of unique labels with varying class numbers. Universally agreed airline standards to identify and safely transport dangerous goods by air. Used and recognised by all major carriers and forwarders. Printed on self-adhesive vinyl tape, with clear polypropylene over-laminate.
  • Corrosive
  • Self-adhesive vinyl
  • Dimensions (L x H) : 100mm x 100mm
  • Class 8