missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Class 3.1 Independent Temperature Safety Device

Product Code. 16424099 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers FED Classic
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16424099

Brand: BINDER™ 80121148

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Class 3.1 Independent Temperature Safety Device with visual alarm (DIN 12880),

  • Class 3.1 Independent Temperature Safety Device
  • For use with assorted BINDER™ Drying and Heating Chambers
  • Features a visual alarm
  • DIN 12880 compliant

Specifications

Certifications/Compliance DIN 12880
Description Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
For Use With (Equipment) BINDER Chambers FED Classic
Type Independent Temperature Safety Device (Class 3.1)
Includes Visual Alarm
Voltage 400 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.