Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
The A1B1E1K1P3 cartridge is part of the wide range of cartridges offered to protect against many hazardous gases, vapors and/or particulates in many applications. The range provides high quality and a cost-effective protection.
Brand:  Honeywell™ N06575089L
Additional Details : Weight : 0.23400kg
Product Code. 15876635
  • £33.71 / 1 pair
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • Protects against organic vapor, acid gas, ammonia and particulates
  • Excellent filtration efficiency over extensive period of time
  • Lower profile and lightweight for optimal comfort
  • Low profile does not obstruct the field of vision
  • Threaded connector is easy to attach
Specifications
Specifications
Class 1 plastic cartridge
Protects against organic vapor, acid gas, ammonia and particulates
122.3 g
EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259
1 Pair