missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ N7700 Class 1 Half-Mask Respirator

Product Code. 15856845
Change view
Click to view available options
Size:
Medium
Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15856845 Medium Each
15866845 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15856845 Supplier Honeywell™ Supplier No. N65770032
  • ONLINE EXCLUSIVE PRICE
    £49.77
    £25.81 / Pack of 1
    Save £23.96
    48% Off
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-performance, half-mask respirator with a lightweight, low-profile design.

  • 100% Silicone hypoallergenic face piece for extra comfort
  • Single unit four-point harness with neck strap is easy to use and its ergonomic design and improved suspension system fits perfectly on top of the head providing a high level of comfort
  • Reduced dead air space means less re-breathed exhaled air
  • Quick to dismantle and clean, with replaceable parts
  • Superior exhalation valve allows more breath to be exhaled so workers feel cooler and more comfortable
  • Variable thickness in sealing area conforms to nose bridge for improved comfort and fit
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC Category PPE: 3, EC Certificate: 250, EC Attestation, EN 140
Color Black
Material Silicone skirt
Product Type Half-mask Respirator
Size Medium
For Use With (Application) Suitable for many types of applications
Harness Style 4-point
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.