missing translation for 'onlineSavingsMsg'
Learn More

Envair™ Class 1 Microbiological Safety Cabinet Model Bio 1+ D

Product Code. 15593306
Change view
Click to view available options
Width (Metric):
900 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric)
15593306 900 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15593306 Supplier Envair™ Supplier No. 30770BIO1D1.0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides microbiological safety for the handling of biological agents up to and including ACDP Hazard Group 3 with a wide range of features.

  • Designed to meet the performance requirements of BS EN 12469:2000 Biotechnology – performance criteria for microbiological safety cabinets with an aperture protection factor greater than or equal to 10 5
  • Comprehensive instrumentation and alarms with easy to clean membrane control panel
  • Available with recirculating, duct and manifold exhaust terminations
  • Clean, attractive stainless steel construction, white polyester coated (outside only)
  • Smooth internal and external surfaces and radiused internal corners for easy cleaning
  • Facility to integrate fumigation systems
  • Designed to exhaust HEPA filtered air to atmosphere via a dedicated ducting system and remote mounted extract fan
  • Integral formalin vaporizer with auto timer
  • Auto-purge to allow full automatic fumigation cycle

Specifications

Product Type Safety Cabinet
Power Consumption 1800 W
Exhaust Air Filter Single Exhaust HEPA Filter
Filter Efficiency Exhaust Rate 0.174 m³/s
For Use With (Application) Handling of Biological Agents
Housing Material Stainless Steel
Dimension (English) Exterior 1.0 m
Width (Metric) 900 mm
Weight (Metric) 136 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.