missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claspin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23206-1-AP-150UL
This item is not returnable.
View return policy
Description
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.Specifications
| Claspin | |
| Polyclonal | |
| Unconjugated | |
| CLSPN | |
| CLASPIN, CLSPN, hClaspin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 63967 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9HAW4 | |
| CLSPN | |
| Claspin Fusion Protein Ag19637 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction