missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claspin Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-248AF594
This item is not returnable.
View return policy
Description
Claspin Polyclonal antibody specifically detects Claspin in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Claspin | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| claspin, claspin homolog, claspin homolog (Xenopus laevis), hClaspin, MGC131612, MGC131613, MGC131615 | |
| The immunogen recognized by this antibody maps to a region between residues 900 and 950 of human Claspin using the numbering given in entry AAG24515.1 (GeneID 63967). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, DNA Repair | |
| 63967 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction