missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLASP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CLASP1 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16830385
|
Proteintech
20803-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16820385
|
Proteintech
20803-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.Specifications
| CLASP1 | |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q7Z460, Q80TV8 | |
| 23332, 304740, 76707 | |
| CLASP1 Fusion Protein Ag14724 | |
| Primary | |
| -20°C | |
| CLASP1 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CLASP1, CLIP associating protein 1, hOrbit1, KIAA0622, MAST1, Multiple asters homolog 1, Protein Orbit homolog 1 | |
| Clasp1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title