missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLASP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | CLASP1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687285
|
Novus Biologicals
NBP2-39049-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18168527
|
Novus Biologicals
NBP2-39049 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLASP1 Polyclonal specifically detects CLASP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CLASP1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CLIP-associating protein 1, cytoplasmic linker associated protein 1, Cytoplasmic linker-associated protein 1, DKFZp686D1968, DKFZp686H2039, hOrbit1, KIAA0622FLJ33821, MAST1FLJ41222, MGC131895, multiple asters 1, Multiple asters homolog 1, Protein Orbit homolog 1 | |
| CLASP1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q7Z460 | |
| 23332 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TSPTNCSHGGLSPSRLWGWSADGLAKHPPPFSQPNSIPTAPSHKALRRSYSPSMLDYDT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title