missing translation for 'onlineSavingsMsg'
Learn More

Clarithromycin, MedChemExpress

Product Code. p-200099037
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16491266

Brand: Medchem Express HY17508

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clarithromycin has a broad spectrum of antimicrobial activity. Clarithromycin inhibits the CYP3A4-catalyzed triazolam alpha-hydroxylation with the IC50 (Ki) value of 56 (43) μM. Clarithromycin significantly inhibits the HERG potassium current.Clarithromycin affects the autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.

Clarithromycin has a broad spectrum of antimicrobial activity. Clarithromycin inhibits the CYP3A4-catalyzed triazolam alpha-hydroxylation with the IC50 (Ki) value of 56 (43) μM. Clarithromycin significantly inhibits the HERG potassium current.Clarithromycin affects the autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.

Specifications

Chemical Name or Material Clarithromycin
Physical Form Solid
CAS 81103-11-9
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C38H69NO13
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 33.33 mg/mL (44.56 mM; Need ultrasonic)
SMILES C[C@@](OC)(C[C@H](C([C@@H]1C)=O)C)[C@@H]([C@H]([C@@H]([C@H](C(O[C@H](CC)[C@@](C)(O)[C@@H]1O)=O)C)O[C@@](O[C@@H](C)[C@@H]2O)([H])C[C@@]2(C)OC)C)O[C@@](O[C@H](C)C[C@@H]3N(C)C)([H])[C@@H]3O
Molecular Weight (g/mol) 747.95
Formula Weight 747.95
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.