missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element Solution For ICP, SPEX CertiPrep™

Product Code. 15668275 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
7 x 125 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15668275 7 x 125 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15668275 Supplier SPEX Certiprep Supplier No. CLMSSETN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Multi-Element Solutions Standards Set for ICP-MS, 125 mL, Set of 7, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • 125 mL Volume
  • Standards Set for ICP-MS

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Multi-Element Solutions without Mercury
Name Note For ICP/MS; Without mercury
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Claritas PPT Grade ASTM I Water Blank,Claritas PPT Grade Hydrochloric Acid Blank,Claritas PPT Grade Nitric Acid Blank
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Element Water, Hydrochloric Acid, Nitric Acid
Solvent or Matrix H2O; 2% HCl; 2% HNO3; 5% HNO3; 10% HCl/1% HNO3; H2O/Tr. HF/Tr. HNO3
Quantity 7 x 125 mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Set of Multi-element Solutions with Mercury
Signal Word
  • Danger||||Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid, hydrochloric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.