Learn More
Memory Test Standard 2 For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CL-MEM-2
Additional Details : CAS Number : 14797-71-8 Weight : 0.12500kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Cl- | 7200μg/mL |
C | 2000μg/mL |
P | 1000μg/mL |
S | 1000μg/mL |
Mo | 20μg/mL |
Sb | 20μg/mL |
Ti | 20μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Memory Test 2 | |
PPT | |
Cl- (7200mg/L),C (2000mg/L),P (1000mg/L) | |
125 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For ICP/MS | |
Triple-leached LDPE bottle | |
H2O/Tr. HF | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Product intended for laboratory use only