missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Claristep™ Station and Tray

Product Code. 15539714 Shop All Sartorius Products
Change view
Click to view available options
Capacity (Metric):
600 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15539714 600 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15539714 Supplier Sartorius Supplier No. 17CM8
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Claristep™ Station and Tray enables the processing of Claristep™ Filters. No vacuum source needed. No power supply needed.

  • Multiplex up to 8 samples
  • Simply position the sample vials in the tray of the station and place the Claristep™ Filters on top
  • After the sample has been filled into the Claristep™ Filter reservoir the station lid is closed by hand and the liquid is filtered
  • White
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 600 μL
Kit Contents Single Tray and Station
Content And Storage Room Temperature
Quantity 1 Ea.
For Use With (Application) HPLC
Target Analytical Sample Preparation Clarification
Type Tools and Accessories
Sufficient For Protein Purification
Format 8-well Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.